missing translation for 'onlineSavingsMsg'
Learn More

3-Thiophenecarbonitrile, 94%, Thermo Scientific™

Product Code. 10711521 Shop All Thermo Scientific Chemicals Products
Click to view available options
Quantity:
1 g
Packaging:
Glass bottle
Unit Size:
1g
This item is not returnable. View return policy

Product Code. 10711521

Brand: Thermo Scientific Acros 263160010

Request Bulk or Custom Formats

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

Specifications

CAS 1641-09-4
CAS Min % 92.0
CAS Max % 100.0
Color Colorless
Boiling Point 82°C (10.0 mmHg)
Infrared Spectrum Authentic
Assay Percent Range 94%
Packaging Glass bottle
Molecular Formula C5H3NS
Refractive Index 1.5595 to 1.5615
MDL Number MFCD00151852
Quantity 1 g
Synonym 3-cyanothiophene, 3-thiophenecarbonitrile, thiophene-3-nitrile, pubchem5508, pubchem12351, acmc-209dqk, ksc176g4h
InChI Key GSXCEVHRIVLFJV-UHFFFAOYSA-N
SMILES C1=CSC=C1C#N
IUPAC Name thiophene-3-carbonitrile
Molecular Weight (g/mol) 109.15
PubChem CID 74231
Formula Weight 109.15
Percent Purity 94%
Physical Form Liquid
Chemical Name or Material 3-Thiophenecarbonitrile, 94%
Show More Show Less
compliance-icons
Product Identifier
  • 3-Thiophenecarbonitrile
Signal Word
  • Warning
Hazard Category
  • Acute toxicity Category 4
Hazard Statement
  • H312-Harmful in contact with skin.
  • H332-Harmful if inhaled.
  • H302-Harmful if swallowed.
Precautionary Statement
  • P280-Wear protective gloves/protective clothing/eye protection/face protection.
  • P301+P312-IF SWALLOWED: Call a POISON CENTER or doctor/physician/ if you feel unwell.
  • P302+P352-IF ON SKIN: Wash with plenty of water/.
  • P304+P340-IF INHALED: Remove person to fresh air and keep comfortable for breathing.

RUO – Research Use Only

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.