missing translation for 'onlineSavingsMsg'
Learn More

3-Thiopheneboronic acid, 97%

Product Code. 10623942 Shop All Thermo Scientific Chemicals Products
Change view
Click to view available options
Quantity:
1 g
5 g
Packaging:
Glass bottle
Unit Size:
1g
5g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Packaging unitSize
10623942 5 g Glass bottle 5g
10051961 1 g Glass bottle 1g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Product Code. 10623942 Supplier Thermo Scientific Acros Supplier No. 345660050
 Restricted Item
In Stock
Add to basket
This item is not returnable. View return policy

CAS: 6165-69-1 | C4H5BO2S | 127.95 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Chemische Identifikatoren

CAS 6165-69-1
Molecular Formula C4H5BO2S
Molecular Weight (g/mol) 127.95
MDL Number MFCD00151851
InChI Key QNMBSXGYAQZCTN-UHFFFAOYSA-N
Synonym 3-thiopheneboronic acid, thiophene-3-boronic acid, 3-thienylboronic acid, thiophene 3-boronic acid, thiophene-3-boronicacid, thiophen-3-yl-3-boronic acid, 3-thienyl boronic acid, 3-thiophene boronic acid, thiophen-3-yl-boranediol, boronic acid, 3-thienyl
PubChem CID 581760
IUPAC Name thiophen-3-ylboronic acid
SMILES OB(O)C1=CSC=C1

Spezifikation

CAS Min % 96.0
CAS Max % 100.0
Melting Point 162°C to 167°C
Color Gray to White
Infrared Spectrum Authentic
Assay Percent Range 97%
Packaging Glass bottle
Quantity 5 g
Solubility Information Solubility in water: .. Other solubilities: soluble in methanol
Formula Weight 127.96
Percent Purity 97%
Physical Form Powder
Chemical Name or Material 3-Thiopheneboronic acid, 97%
Mehr anzeigen Weniger anzeigen
compliance-icons
Produktidentifikator
  • 3-Thiopheneboronic acid
Signalwort
  • Warning
Gefahrenkategorie
  • Serious eye damage/eye irritation Category 2
  • Skin corrosion/irritation Category 2
  • Specific target organ toxicity Category 3
Gefahrenhinweis
  • H319-Causes serious eye irritation.
  • H335-May cause respiratory irritation.
  • H315-Causes skin irritation.
Sicherheitshinweis
  • P261-Avoid breathing dust/fume/gas/mist/vapours/spray.
  • P280-Wear protective gloves/protective clothing/eye protection/face protection.
  • P302+P352-IF ON SKIN: Wash with plenty of water/.
  • P305+P351+P338-IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.

RUO – Research Use Only

Name des Produkts
Wählen Sie ein Thema

Indem Sie auf Absenden klicken, erklären Sie sich damit einverstanden, dass Fisher Scientific sich mit Ihnen in Verbindung setzen kann, um Ihr Feedback in diesem Formular zu bearbeiten. Wir werden Ihre Informationen nicht für andere Zwecke weitergeben. Alle bereitgestellten Kontaktinformationen werden in Übereinstimmung mit unserer Datenschutzrichtlinie aufbewahrt. Datenschutzrichtlinie.