missing translation for 'onlineSavingsMsg'
Learn More

3-Thiopheneacetic acid, 98%

Product Code. 10216980 Shop All Thermo Scientific Chemicals Products
Change view
Click to view available options
Quantity:
10 g
50 g
Packaging:
Glass bottle
Unit Size:
10g
50g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Packaging unitSize
10216980 10 g Glass bottle 10g
10246260 50 g Glass bottle 50g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Product Code. 10216980 Supplier Thermo Scientific Acros Supplier No. 205370100
Request Bulk or Custom Formats

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

CAS: 6964-21-2 | C6H6O2S | 142.17 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 6964-21-2
Molecular Formula C6H6O2S
Molecular Weight (g/mol) 142.17
MDL Number MFCD00005473
InChI Key RCNOGGGBSSVMAS-UHFFFAOYSA-N
Synonym 3-thiopheneacetic acid, 3-thienylacetic acid, 2-thiophen-3-yl acetic acid, thiophene-3-acetic acid, thiophen-3-yl-acetic acid, 3-thiophenacetic acid, thien-3-ylacetic acid, 3-thiopheneethanoic acid, thiophen-3-ylacetic acid, 2-3-thienyl acetic acid
PubChem CID 23404
IUPAC Name 2-thiophen-3-ylacetic acid
SMILES C1=CSC=C1CC(=O)O

Specifications

CAS Min % 97.5
CAS Max % 100.0
Melting Point 77°C to 80°C
Color White to Beige
Infrared Spectrum Authentic
Assay Percent Range 98%
Packaging Glass bottle
Quantity 10 g
Beilstein 18, IV, 4066
Formula Weight 142.17
Percent Purity 98%
Physical Form Crystalline Flakes or Powder
Chemical Name or Material 3-Thiopheneacetic acid, 98%
Show More Show Less
compliance-icons
Product Identifier
  • 3-Thiopheneacetic acid
Signal Word
  • Warning
Hazard Category
  • Serious eye damage/eye irritation Category 2
  • Skin corrosion/irritation Category 2
  • Specific target organ toxicity Category 3
Hazard Statement
  • H319-Causes serious eye irritation.
  • H335-May cause respiratory irritation.
  • H315-Causes skin irritation.
Precautionary Statement
  • P280-Wear protective gloves/protective clothing/eye protection/face protection.
  • P305+P351+P338-IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.

RUO – Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.