missing translation for 'onlineSavingsMsg'
Learn More

3-Thiopheneacetic acid, 98%

Product Code. 11483464
Change view
Click to view available options
Quantity:
5 g
25 g
100 g
Unit Size:
100g
25g
5g
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
11483464 25 g 25g
11473464 5 g 5g
11493464 100 g 100g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
This item is not returnable. View return policy
Product Code. 11483464 Supplier Thermo Scientific Alfa Aesar Supplier No. A10700.14
Request Bulk or Custom Formats

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

CAS: 6964-21-2 | C6H6O2S | 142.172 g/mol

3-Thiopheneacetic acid was used in one-step, size control synthesis of gold nanoparticles. It was also used in the preparation of gold nanoparticles capped with 3-thiopheneacetic acid (3-TAA) via borohydride reduction.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
3-Thiopheneacetic acid was used in one-step, size control synthesis of gold nanoparticles. It was also used in the preparation of gold nanoparticles capped with 3-thiopheneacetic acid (3-TAA) via borohydride reduction.

Solubility
Very soluble in water.

Notes
Stable under recommended storage conditions. Incompatible with oxidizing agents and bases.
TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 6964-21-2
Molecular Formula C6H6O2S
Molecular Weight (g/mol) 142.172
MDL Number MFCD00005473
InChI Key RCNOGGGBSSVMAS-UHFFFAOYSA-N
Synonym 3-thiopheneacetic acid, 3-thienylacetic acid, 2-thiophen-3-yl acetic acid, thiophene-3-acetic acid, thiophen-3-yl-acetic acid, 3-thiophenacetic acid, thien-3-ylacetic acid, 3-thiopheneethanoic acid, thiophen-3-ylacetic acid, 2-3-thienyl acetic acid
PubChem CID 23404
IUPAC Name 2-thiophen-3-ylacetic acid
SMILES C1=CSC=C1CC(=O)O

Specifications

Melting Point 77°C to 81°C
Boiling Point 129°C (2.5 mmHg)
Quantity 25 g
UN Number UN3261
Beilstein 113622
Solubility Information Very soluble in water.
Formula Weight 142.18
Percent Purity 98%
Chemical Name or Material 3-Thiopheneacetic acid

RUO – Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.