missing translation for 'onlineSavingsMsg'
Learn More

3-(Thiophen-2-yl)propanoic Acid, Mikromol

Product Code. 30463973
Change view
Click to view available options
Quantity:
25 mg
4 x 25 mg
Unit Size:
100mg
25mg
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
30463973 4 x 25 mg 100mg
30050828 25 mg 25mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Product Code. 30463973 Supplier Mikromol Supplier No. MM1577.0200254AMP

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

Discover Mikromol- your trusted source for high-quality pharmaceutical reference standards. Supporting accurate, compliant analysis with ISO 17034-accredited materials designed for confidence in every result.

3-(Thiophen-2-yl)propanoic Acid, Mikromol
TRUSTED_SUSTAINABILITY

Specifications

Chemical Name or Material 3-(Thiophen-2-yl)propanoic Acid
CAS 5928-51-8
Analyte or Component Names 3-(Thiophen-2-yl)propanoic Acid
Shipping Condition Room Temperature
Molecular Formula C7 H8 O2 S
Certifications/Compliance ISO 17025
Synonym 2-Thiophenepropanoic acid, 2-Thiophenepropionic acid (6CI,7CI,8CI), 3-(2-Thienyl)propanoic acid, 3-(2-Thienyl)propionic acid, 3-(Thiophen-2-yl)propanoic acid, 3-(Thiophen-2-yl)propionic acid, beta-(2-Thienyl)propionic acid, Eprosartan Related Compound B
InChI Formula InChI=1S/C7H8O2S/c8-7(9)4-3-6-2-1-5-10-6/h1-2,5H,3-4H2,(H,8,9)
Solution Type Neat
SMILES OC(=O)CCc1cccs1
IUPAC Name 3-thiophen-2-ylpropanoic acid
Molecular Weight (g/mol) 156.2
Quantity 4 x 25 mg
Formula Weight 156.0245 g/mol
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.