missing translation for 'onlineSavingsMsg'
Learn More

3-Thienylmethylamine, 97%, Thermo Scientific™

Product Code. 10011744
Change view
Click to view available options
Quantity:
250 mg
1 g
5 g
Packaging:
Amber Glass Bottle
Unit Size:
1g
250mg
5g
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Packaging unitSize
10011744 5 g Amber Glass Bottle 5g
10334232 250 mg Amber Glass Bottle 250mg
10394322 1 g Amber Glass Bottle 1g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
This item is not returnable. View return policy
Product Code. 10011744 Supplier Thermo Scientific Maybridge Supplier No. CC13113DE
Request Bulk or Custom Formats

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

Chemical Identifiers

CAS 27757-86-4
Molecular Formula C5H7NS
Molecular Weight (g/mol) 113.178
MDL Number MFCD01529872
InChI Key DUDAKCCDHRNMDJ-UHFFFAOYSA-N
Synonym 3-thienylmethylamine, 3-aminomethyl thiophene, 3-thiophenemethanamine, 3-thiophenemethylamine, 1-thien-3-ylmethanamine, 3-thienylmethyl amine, 1-3-thienyl methanamine, thiophen-3-yl-methylamine, thiophen-3-yl methanamine, thien-3-yl methylamine
PubChem CID 2776381
IUPAC Name thiophen-3-ylmethanamine
SMILES C1=CSC=C1CN

Specifications

CAS Min % 96.5
CAS Max % 100.0
Assay Percent Range 97%
Packaging Amber Glass Bottle
Quantity 5 g
Formula Weight 113.18
Percent Purity 97%
Chemical Name or Material 3-Thienylmethylamine
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.