missing translation for 'onlineSavingsMsg'
Learn More

3-Thienylboronic acid, 97%, Maybridge™

Product Code. 10118454
Change view
Click to view available options
Quantity:
1 g
5 g
Unit Size:
1g
5g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
10118454 5 g 5g
10001744 1 g 1g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Product Code. 10118454 Supplier Thermo Scientific Maybridge Supplier No. CC13112DE
Request Bulk or Custom Formats

Please to purchase this item. Need a web account? Register with us today!

This item is currently unavailable or has been discontinued.
View the product page for possible alternatives.
View alternative products

This item is not returnable. View return policy

Chemical Identifiers

CAS 6165-69-1
Molecular Formula C4H5BO2S
Molecular Weight (g/mol) 127.95
MDL Number MFCD00151851
InChI Key QNMBSXGYAQZCTN-UHFFFAOYSA-N
Synonym 3-thiopheneboronic acid, thiophene-3-boronic acid, 3-thienylboronic acid, thiophene 3-boronic acid, thiophene-3-boronicacid, thiophen-3-yl-3-boronic acid, 3-thienyl boronic acid, 3-thiophene boronic acid, thiophen-3-yl-boranediol, boronic acid, 3-thienyl
PubChem CID 581760
IUPAC Name (thiophen-3-yl)boronic acid
SMILES OB(O)C1=CSC=C1

Specifications

Assay Percent Range 97%
Quantity 5 g
Formula Weight 127.96
Percent Purity 97%
Chemical Name or Material 3-Thienylboronic acid
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.