missing translation for 'onlineSavingsMsg'
Learn More

(3-Thien-3-ylphenyl)methanol, ≥97%, Thermo Scientific™

Product Code. 10301153
Change view
Click to view available options
Quantity:
250 mg
1 g
5 g
Unit Size:
1g
250mg
5g
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
10301153 1 g 1g
10341103 250 mg 250mg
10643055 5 g 5g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
This item is not returnable. View return policy
Product Code. 10301153 Supplier Thermo Scientific Maybridge Supplier No. CC48409DA
Request Bulk or Custom Formats

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

Chemical Identifiers

CAS 89929-82-8
Molecular Formula C11H10OS
Molecular Weight (g/mol) 190.26
MDL Number MFCD06203083
InChI Key VSWBZMMHNMDWJL-UHFFFAOYSA-N
Synonym 3-thien-3-ylphenyl methanol, 3-thiophen-3-yl phenyl methanol, 3-3-thienyl phenyl methanol, 3-thiophen-3-ylphenyl methanol, acmc-20ls08, 3-3-thienyl phenylmethanol, 3-3-thiophenyl phenyl methanol, 3-3-thienyl phenyl methan-1-ol
PubChem CID 7164662
IUPAC Name (3-thiophen-3-ylphenyl)methanol
SMILES C1=CC(=CC(=C1)CO)C2=CSC=C2

Specifications

CAS Min % 94.5
CAS Max % 100.0
Assay Percent Range 95%
Packaging Amber glass bottle
Quantity 1 g
Formula Weight 190.27
Percent Purity 95%
Chemical Name or Material (3-Thien-3-ylphenyl)methanol
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.