missing translation for 'onlineSavingsMsg'
Learn More

(3-Thien-2-ylphenyl)methanol, ≥97%, Thermo Scientific™

Product Code. p-3176111
Click to view available options
Quantity:
250 mg
1 g
5 g
Unit Size:
1g
250mg
5g
This item is not returnable. View return policy

Product Code. 10387594

Brand: Thermo Scientific Maybridge CC44509CB

Request Bulk or Custom Formats

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

Chemical Identifiers

CAS 103669-00-7
Molecular Formula C11H10OS
Molecular Weight (g/mol) 190.26
MDL Number MFCD06203108
InChI Key SATFADRDDGOLJP-UHFFFAOYSA-N
Synonym 3-thien-2-ylphenyl methanol, 3-2-thienyl benzyl alcohol, 3-thiophen-2-yl phenyl methanol, 3-2-thienyl phenyl methanol, benzenemethanol,3-2-thienyl, acmc-1c9kv, 3-thiophen-2-ylphenyl methanol, 3-thiophen-2-yl benzyl alcohol, 3-2-thienyl phenyl methan-1-ol
PubChem CID 7537586
SMILES OCC1=CC=CC(=C1)C1=CC=CS1

Specifications

CAS Min % 96.5
CAS Max % 100.0
Assay Percent Range 97%
Packaging Amber glass bottle
Quantity 250 mg
IUPAC Name [3-(thiophen-2-yl)phenyl]methanol
Formula Weight 190.27
Percent Purity 97%
Chemical Name or Material (3-Thien-2-ylphenyl)methanol
Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.