missing translation for 'onlineSavingsMsg'
Learn More

(3-Thien-2-ylphenyl)methanol, ≥97%, Thermo Scientific™

Product Code. 10614595
Change view
Click to view available options
Quantity:
250 mg
1 g
5 g
Unit Size:
1g
250mg
5g
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Unit Size
10614595 1 g 1g
10387594 250 mg 250mg
10352105 5 g 5g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
This item is not returnable. View return policy
Product Code. 10614595 Supplier Thermo Scientific Maybridge Supplier No. CC44509DA
 Restricted Item
Estimated Shipment: 08-12-2026
Add to basket

missing translation for 'validMainframeMsg'

This item is not returnable. View return policy

Chemical Identifiers

CAS 103669-00-7
Molecular Formula C11H10OS
Molecular Weight (g/mol) 190.26
MDL Number MFCD06203108
InChI Key SATFADRDDGOLJP-UHFFFAOYSA-N
Synonym 3-thien-2-ylphenyl methanol, 3-2-thienyl benzyl alcohol, 3-thiophen-2-yl phenyl methanol, 3-2-thienyl phenyl methanol, benzenemethanol,3-2-thienyl, acmc-1c9kv, 3-thiophen-2-ylphenyl methanol, 3-thiophen-2-yl benzyl alcohol, 3-2-thienyl phenyl methan-1-ol
PubChem CID 7537586
SMILES OCC1=CC=CC(=C1)C1=CC=CS1

Specifications

CAS Min % 96.5
CAS Max % 100.0
Assay Percent Range 97%
Packaging Amber glass bottle
Quantity 1 g
IUPAC Name (3-thiophen-2-ylphenyl)methanol
Formula Weight 190.27
Percent Purity 97%
Chemical Name or Material (3-Thien-2-ylphenyl)methanol
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.