missing translation for 'onlineSavingsMsg'
Learn More

3-Thien-2-ylbenzonitrile 97+%, Thermo Scientific™

Product Code. 10644405
Change view
Click to view available options
Quantity:
1 g
Packaging:
Amber Glass Bottle
Unit Size:
1g
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Packaging unitSize
10644405 1 g Amber Glass Bottle 1g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Product Code. 10644405 Supplier Thermo Scientific Maybridge Supplier No. CC44516DA
Request Bulk or Custom Formats

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

Specifications

CAS 380626-35-7
Assay Percent Range 97%
Packaging Amber Glass Bottle
Molecular Formula C11H7NS
MDL Number MFCD02323389
Quantity 1 g
Synonym 3-thien-2-ylbenzonitrile, 3-thiophen-2-yl benzonitrile, 3-2-thienyl benzonitrile, 3-thiophen-2-yl-benzonitrile, 3-2-thienyl benzenecarbonitrile
InChI Key DIILPQRSOQWSCU-UHFFFAOYSA-N
SMILES N#CC1=CC(=CC=C1)C1=CC=CS1
IUPAC Name 3-thiophen-2-ylbenzonitrile
Molecular Weight (g/mol) 185.24
PubChem CID 4645033
Formula Weight 185.25
Percent Purity 97%
Chemical Name or Material 3-Thien-2-ylbenzonitrile
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.