missing translation for 'onlineSavingsMsg'
Learn More

3-tert-Butylphenol, TRC

Product Code. 30345503
Change view
Click to view available options
Quantity:
1 g
Unit Size:
1g
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
30345503 1 g 1g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Product Code. 30345503 Supplier Toronto Research Chemicals Supplier No. TRCB8090801G

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.

TRUSTED_SUSTAINABILITY

Specifikationer

Chemical Name or Material 3-tert.-Butylphenol
CAS 585-34-2
Molecular Formula C10 H14 O
Quantity 1 g
Synonym m-tert-Butylphenol
InChI Formula InChI=1S/C10H14O/c1-10(2,3)8-5-4-6-9(11)7-8/h4-7,11H,1-3H3
SMILES CC(C)(C)c1cccc(O)c1
IUPAC Name 3-tert-butylphenol
Molecular Weight (g/mol) 150.22
Formula Weight 150.1045
Produkttitel
Välj ett problem

Genom att klicka på Skicka bekräftar du att du kan bli kontaktad av Fisher Scientific angående feedbacken du har lämnat i detta formulär. Vi kommer inte att dela din information för andra ändamål. All kontaktinformation som tillhandahålls ska också underhållas i enlighet med vår Sekretesspolicy.