missing translation for 'onlineSavingsMsg'
Learn More

3-tert-Butylphenol, 99+%

Product Code. 10331474 Shop All Thermo Scientific Chemicals Products
Change view
Click to view available options
Quantity:
1 g
25 g
100 g
Packaging:
Glass bottle
Plastic bottle
Unit Size:
100g
1g
25g
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Packaging unitSize
10331474 1 g Glass bottle 1g
10143060 25 g Plastic bottle 25g
10347470 100 g Plastic bottle 100g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
This item is not returnable. View return policy
Product Code. 10331474 Supplier Thermo Scientific Acros Supplier No. 154380010
Request Bulk or Custom Formats

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

CAS: 585-34-2 | C10H14O | 150.22 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 585-34-2
Molecular Formula C10H14O
Molecular Weight (g/mol) 150.22
MDL Number MFCD00002300
InChI Key CYEKUDPFXBLGHH-UHFFFAOYSA-N
Synonym m-tert-butylphenol, phenol, 3-1,1-dimethylethyl, 3-t-butylphenol, 3-tert-butyl phenol, phenol, m-tert-butyl, 3-tert-butyl-phenol, 3-tertbutylphenol, 3-tert.butylphenol, m-tert.-butylphenol
PubChem CID 11450
ChEBI CHEBI:34348
IUPAC Name 3-tert-butylphenol
SMILES CC(C)(C)C1=CC=CC(O)=C1

Specifications

CAS Min % 99.0
CAS Max % 100.0
Melting Point 40°C to 43°C
Color White
Boiling Point 125°C to 130°C (20.0 mmHg)
Flash Point 108°C
Infrared Spectrum Authentic
Assay Percent Range 99% min. (GC)
Packaging Glass bottle
Linear Formula (CH3)3CC6H4OH
Quantity 1 g
Solubility Information Solubility in water: .. Other solubilities: soluble in methanol
Formula Weight 150.22
Percent Purity 99+%
Physical Form Crystalline Powder
Chemical Name or Material 3-tert-Butylphenol, 99+%
Show More Show Less
compliance-icons compliance-icons
Product Identifier
  • 3-tert-Butylphenol
Signal Word
  • Danger
Hazard Category
  • Acute toxicity Category 4
  • Serious eye damage/eye irritation Category 1
  • Skin corrosion/irritation Category 1 B
Hazard Statement
  • H318-Causes serious eye damage.
  • H314-Causes severe skin burns and eye damage.
  • H312-Harmful in contact with skin.
  • H332-Harmful if inhaled.
  • H302-Harmful if swallowed.
Precautionary Statement
  • P280-Wear protective gloves/protective clothing/eye protection/face protection.
  • P301+P312-IF SWALLOWED: Call a POISON CENTER or doctor/physician/ if you feel unwell.
  • P301+P330+P331-IF SWALLOWED: rinse mouth. Do NOT induce vomiting.
  • P302+P352-IF ON SKIN: Wash with plenty of water/.
  • P304+P340-IF INHALED: Remove person to fresh air and keep comfortable for breathing.
  • P305+P351+P338-IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
  • P310-Immediately call a POISON CENTER/doctor/.

RUO – Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.