missing translation for 'onlineSavingsMsg'
Learn More
Learn More
3-[(tert-Butyldimethylsilyl)oxy]-1-propanol, TRC
Click to view available options
Quantity:
1 g
Unit Size:
1g
Specifications
Specifications
| Chemical Name or Material | 3-[(tert-Butyldimethylsilyl)oxy]-1-propanol |
| CAS | 73842-99-6 |
| Molecular Formula | C9 H22 O2 Si |
| Quantity | 1 g |
| InChI Formula | InChI=1S/C9H22O2Si/c1-9(2,3)12(4,5)11-8-6-7-10/h10H,6-8H2,1-5H3 |
| SMILES | CC(C)(C)[Si](C)(C)OCCCO |
| IUPAC Name | 3-[tert-butyl(dimethyl)silyl]oxypropan-1-ol |
| Molecular Weight (g/mol) | 190.36 |
| Formula Weight | 190.1389 |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction