missing translation for 'onlineSavingsMsg'
Learn More

3-(tert-Butyldimethylsiloxy)thiophenol, 95%, Thermo Scientific Chemicals

Product Code. 11364006
Change view
Click to view available options
Quantity:
1 g
5 g
Unit Size:
1g
5g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Unit Size
11364006 5 g 5g
11354006 1 g 1g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Product Code. 11364006 Supplier Thermo Scientific Alfa Aesar Supplier No. L17238.06
 Restricted Item
In Stock
Add to basket

missing translation for 'validMainframeMsg'

This item is not returnable. View return policy

CAS: 216393-56-5 | Molecular Formula: C12H20OSSi | Molecular Weight: 240.44 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 216393-56-5
Molecular Formula C12H20OSSi
Molecular Weight (g/mol) 240.44
MDL Number MFCD01318115
InChI Key KRHCTXFJCFMFOR-UHFFFAOYSA-N
Synonym 3-tert-butyldimethylsilyl oxy benzenethiol, 3-tert-butyldimethylsiloxy thiophenol, acmc-20ani3, 3-tert-butyl-dimethylsiloxy-thiophenol, 3-tert-butyldimethylsilyloxy benzenethiol, 3-t-butyl-dimethyl-silanyloxy-benzenethiol, 3-tert-butyl-dimethyl-silanyloxy-benzenethiol, 3-tert-butyl dimethyl silyl oxy benzene-1-thiol, 3-1,1-dimethylethyl dimethyl silyl oxy benzenethiol
PubChem CID 21976868
SMILES CC(C)(C)[Si](C)(C)OC1=CC(S)=CC=C1

Specifications

Refractive Index 1.521
Quantity 5 g
Sensitivity Air Sensitive
IUPAC Name 3-[tert-butyl(dimethyl)silyl]oxybenzenethiol
Formula Weight 240.44
Percent Purity 95%
Chemical Name or Material 3-(tert-Butyldimethylsiloxy)thiophenol

RUO – Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.