missing translation for 'onlineSavingsMsg'
Learn More

3-(tert-Butyldimethylsiloxy)glutaric anhydride, 95%

Product Code. 15496366
Click to view available options
Quantity:
5 g
25 g
Unit Size:
25g
5g
This item is not returnable. View return policy

Product Code. 15496366

Brand: Thermo Scientific Alfa Aesar H61227.14

Request Bulk or Custom Formats

Please to purchase this item. Need a web account? Register with us today!

This item is currently unavailable or has been discontinued.
View the product page for possible alternatives.
View alternative products

This item is not returnable. View return policy

CAS: 91424-40-7 | C11H20O4Si | 244.362 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 91424-40-7
Molecular Formula C11H20O4Si
Molecular Weight (g/mol) 244.362
MDL Number MFCD00075235
InChI Key RXAJGRHLLRGVSB-UHFFFAOYSA-N
Synonym 3-tert-butyldimethylsilyloxy glutaric anhydride, 4-tert-butyldimethylsilyl oxy oxane-2,6-dione, 3-tert-butyldimethylsilyl oxy pentanedioic anhydride, 3-tert-butyldimethylsilyloxy glutaric anhydride,, 4-tert-butyldimethylsilyl oxy dihydro-2h-pyran-2,6 3h-dione, acmc-209rbg, 4-tert-butyl dimethyl silyl oxyoxane-2,6-dione, 3-tert-butyldimethylsilyloxy glutaricanhydride, 3-tert-butyldimethylsiloxy pentanedioic anhydride, 3-tert-butyldimethyl silyl oxy glutaric anhydride
PubChem CID 4141040
IUPAC Name 4-[tert-butyl(dimethyl)silyl]oxyoxane-2,6-dione
SMILES CC(C)(C)[Si](C)(C)OC1CC(=O)OC(=O)C1

Specifications

Melting Point 79°C to 81°C
Density 1.030 g/mL
Flash Point >113°C (235°F)
Refractive Index 1.451
Quantity 25 g
Sensitivity Moisture sensitive
Formula Weight 244.36
Percent Purity 98%
Physical Form Solid
Chemical Name or Material 3-(tert-Butyldimethylsiloxy)glutaric anhydride

RUO – Research Use Only

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.