missing translation for 'onlineSavingsMsg'
Learn More
Learn More
3-tert-Butyl-p-hydroxyanisole, TRC
Click to view available options
Quantity:
100 g
Unit Size:
100g
Specifications
Specifications
| Chemical Name or Material | 3-tert-Butyl-4-hydroxyanisole |
| CAS | 121-00-6 |
| Molecular Formula | C11 H16 O2 |
| Quantity | 100 g |
| Synonym | Phenol, 2-tert-butyl-4-methoxy- (8CI), 2-(1,1-Dimethylethyl)-4-methoxyphenol, 2-tert-Butyl-4-methoxyphenol, 2-tert-Butyl-p-methoxyphenol, 3-(1,1-Dimethylethyl)-4-hydroxyanisole, 3-BHA, 3-tert-Butyl-4-hydroxyanisole, 3-tert-Butyl-p-hydroxyanisole, 4-Hydroxy-3-tert-butylanisole, 4-Methoxy-2-tert-butylphenol, 4-Methoxy-6-tert-butylphenol, o-tert-Butyl-p-methoxyphenol, tert-Butyl-2-methoxy-4-phenol |
| InChI Formula | InChI=1S/C11H16O2/c1-11(2,3)9-7-8(13-4)5-6-10(9)12/h5-7,12H,1-4H3 |
| SMILES | COc1ccc(O)c(c1)C(C)(C)C |
| IUPAC Name | 2-tert-butyl-4-methoxyphenol |
| Molecular Weight (g/mol) | 180.24 |
| Formula Weight | 180.115 |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction