missing translation for 'onlineSavingsMsg'
Learn More

3-(tert-Butyl)aniline, 97%, Maybridge™

Product Code. 10140742
Change view
Click to view available options
Quantity:
250 mg
1 g
Packaging:
Amber Glass Bottle
Unit Size:
1g
250mg
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Packaging Unit Size
10140742 250 mg Amber Glass Bottle 250mg
10019301 1 g Amber Glass Bottle 1g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Product Code. 10140742 Supplier Thermo Scientific Maybridge Supplier No. RF01575CB

This item is currently unavailable or has been discontinued.
View the product page for possible alternatives.
View alternative products

This item is not returnable. View return policy

Chemical Identifiers

CAS 5369-19-7
Molecular Formula C10H15N
Molecular Weight (g/mol) 149.24
MDL Number MFCD00125078
InChI Key DPKTVUKEPNBABS-UHFFFAOYSA-N
Synonym 3-tert-butyl aniline, 3-tert-butylbenzenamine, benzenamine, 3-1,1-dimethylethyl, 3-t-butyl aniline, 3-tert-butyl-phenylamine, 3-tert-butyl phenylamine, 3-tert-butylphenyl amine, m-t-butylaniline, 3-t-butylaniline, m-tert-butylaniline
PubChem CID 79334
IUPAC Name 3-tert-butylaniline
SMILES CC(C)(C)C1=CC=CC(N)=C1

Specifications

CAS Min % 97.0
Assay Percent Range 97%
Packaging Amber Glass Bottle
Linear Formula (CH3)3CC6H4NH2
Quantity 250 mg
Formula Weight 149.24
Percent Purity 97%
Chemical Name or Material 3-(Tert-butyl)aniline
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.