missing translation for 'onlineSavingsMsg'
Learn More

3-tert-Butyl-5-chloro-6-hydroxymethyluracil (6-Desmethyl-6-Hydroxymethyl Terbacil), TRC

Product Code. 30180112
Change view
Click to view available options
Quantity:
10 mg
50 mg
100 mg
Unit Size:
100mg
10mg
50mg
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Unit Size
30180112 50 mg 50mg
30179727 10 mg 10mg
30180094 100 mg 100mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
This item is not returnable. View return policy
Product Code. 30180112 Supplier Toronto Research Chemicals Supplier No. TRCB99709550MG
 Restricted Item
Estimated Shipment: 27-10-2026
Add to basket
Add to basket

missing translation for 'validMainframeMsg'

This item is not returnable. View return policy

High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 25546-02-5
Molecular Formula C9 H13 Cl N2 O3
Molecular Weight (g/mol) 232.6641
Synonym Uracil, 3-tert-butyl-5-chloro-6-(hydroxymethyl)- (8CI), 5-Chloro-3-(1,1-dimethylethyl)-6-(hydroxymethyl)-2,4(1H,3H)-pyrimidinedione, 3-tert-Butyl-5-chloro-6-hydroxymethyluracil
IUPAC Name 3-tert-butyl-5-chloro-6-(hydroxymethyl)-1H-pyrimidine-2,4-dione
SMILES CC(C)(C)N1C(=O)NC(=C(Cl)C1=O)CO

Specifications

Quantity 50 mg
InChI Formula InChI=1S/C9H13ClN2O3/c1-9(2,3)12-7(14)6(10)5(4-13)11-8(12)15/h13H,4H2,1-3H3,(H,11,15)
Formula Weight 232.0615
Chemical Name or Material Terbacil-hydroxymethyl
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.