missing translation for 'onlineSavingsMsg'
Learn More

3-tert-Butyl-4-hydroxyanisole, 98%

Product Code. 11984751 Shop All Thermo Scientific Chemicals Products
Change view
Click to view available options
Quantity:
100 g
500 g
Unit Size:
100g
500g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
11984751 500 g 500g
11351473 100 g 100g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Product Code. 11984751 Supplier Thermo Scientific Acros Supplier No. 439695000
Request Bulk or Custom Formats

Please to purchase this item. Need a web account? Register with us today!

This item is currently unavailable or has been discontinued.
View the product page for possible alternatives.
View alternative products

This item is not returnable. View return policy

CAS: 121-00-6 | C11H16O2 | 180.25 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 121-00-6
Molecular Formula C11H16O2
Molecular Weight (g/mol) 180.25
InChI Key MRBKEAMVRSLQPH-UHFFFAOYSA-N
Synonym 3-tert-butyl-4-hydroxyanisole, 4-hydroxy-3-tert-butylanisole, 2-tert-butyl-4-methoxyphenol, 3-bha, 4-methoxy-2-tert-butylphenol, 3-t-butyl-4-hydroxyanisole, phenol, 2-1,1-dimethylethyl-4-methoxy, 2-1,1-dimethylethyl-4-methoxyphenol, 2-butyl-4-hydroxyanisole, phenol, 2-tert-butyl-4-methoxy
PubChem CID 8456
ChEBI CHEBI:76358
IUPAC Name 2-tert-butyl-4-methoxyphenol
SMILES CC(C)(C)C1=C(C=CC(=C1)OC)O

Specifications

CAS Min % 97.5
CAS Max % 100.0
Melting Point 58.0°C to 66.0°C
Color White
Flash Point 110°C
Infrared Spectrum Authentic
Assay Percent Range 97.5% min. (GC)
Quantity 500 g
Solubility Information Solubility in water: in water:(20°C) insoluble.,in ethanol:(20°C) soluble
Formula Weight 180.25
Percent Purity 98%
Physical Form Powder or Waxy Solid
Chemical Name or Material 3-tert-Butyl-4-hydroxyanisole
Show More Show Less
compliance-icons
Product Identifier
  • 3-tert-Butyl-4-hydroxyanisole
Signal Word
  • Warning
Hazard Category
  • Serious eye damage/eye irritation Category 2
  • Respiratory sensitiser Category 1
  • Skin corrosion/irritation Category 2
Hazard Statement
  • H317-May cause an allergic skin reaction.
  • H319-Causes serious eye irritation.
  • H315-Causes skin irritation.
Precautionary Statement
  • P280-Wear protective gloves/protective clothing/eye protection/face protection.
  • P302+P352-IF ON SKIN: Wash with plenty of water/.
  • P305+P351+P338-IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
  • P333+P313-If skin irritation or rash occurs: Get medical advice/attention.

RUO – Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.