missing translation for 'onlineSavingsMsg'
Learn More

3-Sulfolene, 98%

Product Code. p-7021524
Change view
Click to view available options
Quantity:
100 g
500 g
1000 g
5000 g
Unit Size:
1000g
100g
5000g
500g
4 product options available for selection
Product selection table with 4 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
11309459 100 g 100g
11319459 500 g 500g
11431588 1000 g 1000g
11441588 5000 g 5000g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 4 options available.
4 options
This item is not returnable. View return policy
Product Code. 11309459 Supplier Thermo Scientific Alfa Aesar Supplier No. A13887.22
Request Bulk or Custom Formats

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

CAS: 77-79-2 | C4H6O2S | 118.15 g/mol

It is a source of cisoid butadiene for Diels-Alder reactions. This chemical is also a convenient source of sulfur dioxide. It is a solvent used in the petrochemical industry for the extraction of aromatics from hydrocarbon streams.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
It is a source of cisoid butadiene for Diels-Alder reactions. This chemical is also a convenient source of sulfur dioxide. It is a solvent used in the petrochemical industry for the extraction of aromatics from hydrocarbon streams.

Solubility
Soluble in water, ethanol, benzene, ether.

Notes
Store in a cool, dry, well-ventilated area. Store away from oxidizing agent.
TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 77-79-2
Molecular Formula C4H6O2S
Molecular Weight (g/mol) 118.15
MDL Number MFCD00005481
InChI Key MBDNRNMVTZADMQ-UHFFFAOYSA-N
Synonym 3-sulfolene, butadiene sulfone, sulfolene, thiophene, 2,5-dihydro-, 1,1-dioxide, beta-sulfolene, 2,5-dihydrothiophene sulfone, 2,5-dihydrothiophene dioxide, 2,5-dihydrothiophene-1,1-dioxide, sulfol-3-ene, ccris 569
PubChem CID 6498
IUPAC Name 2,5-dihydrothiophene 1,1-dioxide
SMILES C1C=CCS1(=O)=O

Specifications

Melting Point 63°C to 67°C
Density 1.314
Flash Point 112°C (233°F)
Quantity 100 g
Beilstein 107004
Merck Index 14,8956
Solubility Information Soluble in water,ethanol,benzene,ether.
Formula Weight 118.15
Percent Purity 98%
Chemical Name or Material 3-Sulfolene

RUO – Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.