missing translation for 'onlineSavingsMsg'
Learn More

3-Sulfo-N-succinimidyl 4-(maleimidomethyl)cyclohexane-1-carboxylate sodium salt, 97+%

Product Code. 15445275
Click to view available options
Quantity:
25 mg
100 mg
Unit Size:
100mg
25mg
This item is not returnable. View return policy

Product Code. 15445275

Brand: Thermo Scientific Alfa Aesar H54807.MB

Request Bulk or Custom Formats

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

CAS: 92921-24-9 | C16H17N2NaO9S | 436.367 g/mol

3-Sulfo-N-succinimidyl 4-(maleimidomethyl)cyclohexane-1-carboxylate sodium salt is a cross-linking reagent incorporating an extended spacer with amine and sulfhydryl reactivity. Typically, coupled initially to molecules containing primary amines by amide bonds buffered at pH 7.5 (6.5-8.5). Second coupling specific for molecules containing free sulfhydryl by thioether linkage buffered at pH 6.8 (6.5-7.0). Useful for preparation of enzyme immunoconjugates and hapten carrier molecule conjugates.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
3-Sulfo-N-succinimidyl 4-(maleimidomethyl)cyclohexane-1-carboxylate sodium salt is a cross-linking reagent incorporating an extended spacer with amine and sulfhydryl reactivity. Typically, coupled initially to molecules containing primary amines by amide bonds buffered at pH 7.5 (6.5-8.5). Second coupling specific for molecules containing free sulfhydryl by thioether linkage buffered at pH 6.8 (6.5-7.0). Useful for preparation of enzyme immunoconjugates and hapten carrier molecule conjugates.

Solubility
Soluble in water (2.5 mg/ml) and dimethyl formamide.

Notes
Keep container tightly sealed. Store in cool, dry conditions in well sealed containers. Incompatible with oxidizing agents.
TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 92921-24-9
Molecular Formula C16H17N2NaO9S
Molecular Weight (g/mol) 436.367
MDL Number MFCD00054978
InChI Key VUFNRPJNRFOTGK-UHFFFAOYSA-M
Synonym sulfo-smcc, sodium 1-4-2,5-dioxo-2,5-dihydro-1h-pyrrol-1-yl methyl cyclohexanecarbonyl oxy-2,5-dioxopyrrolidine-3-sulfonate, sulfo-smcc sodium, 4-maleimidomethyl cyclohexane-1-carboxylic acid 3-sulfo-n-hydroxysuccinimide ester sodium salt, 3-sulfo-n-succinimidyl 4-n-maleimidomethyl cyclohexane-1-carboxylate sodium salt, sulfo-trans-smcc, sulfo-smcc crosslinker, sulfo-n-succinimidyl 4-maleimidomethyl cyclohexane-1-carboxylate sodium salt, sulfo-smcc sodium salt, bicl207
PubChem CID 4618794
IUPAC Name sodium;1-[4-[(2,5-dioxopyrrol-1-yl)methyl]cyclohexanecarbonyl]oxy-2,5-dioxopyrrolidine-3-sulfonate
SMILES C1CC(CCC1CN2C(=O)C=CC2=O)C(=O)ON3C(=O)CC(C3=O)S(=O)(=O)[O-].[Na+]

Specifications

Quantity 25 mg
Solubility Information Soluble in water (2.5mg/ml) and dimethyl formamide.
Formula Weight 436.37
Percent Purity ≥98%
Chemical Name or Material 3-Sulfo-N-succinimidyl 4-(maleimidomethyl)cyclohexane-1-carboxylate sodium salt

RUO – Research Use Only

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.