missing translation for 'onlineSavingsMsg'
Learn More

Masterflex™ 3-Stop Tubing, Tygon LMT-55

Designed for optimal operation in your Ismatec pump.

Brand:  Masterflex™ 97619-40

Additional Details : Weight : 1.00000kg

 View more versions of this product

Product Code. 15935811

  • £156.00 / Pack of 12
Estimated Shipment: 10-10-2024
to see stock.
Add to basket

Description

Description

  • For use with pumps that incorporate cassettes into the pumphead design.
  • Color-coded stops.
  • Inexpensive all-around tubing for general laboratory applications.
  • Transparent.
  • Resistant to almost all inorganic chemicals.
  • Smooth polished inner wall.
  • Low gas permeability.
  • Non-aging and non-oxidizing.
Specifications

Specifications

Green-Green
Tygon LMT-55
General laboratory applications, pumps with cassettes in pumphead design
Acids: Good, Alkaline solutions: Good, Solvents: Not rec., Pressure: Fair, Vacuum: Good, Viscous media: Excellent, Sterile media: Limited
Transparent
1.85 mm
406.4 mm
12/Pk
I.D. 1.85 mm
Product Suggestions

Product Suggestions

Videos
SDS
Documents

Documents

Certificates
Special Offers

Special Offers