missing translation for 'onlineSavingsMsg'
Learn More

Masterflex™ 3-Stop Tubing, Tygon LMT-55

Designed for optimal operation in your Ismatec pump.

Brand:  Masterflex™ SC0051T

Code : NEW

Additional Details : Weight : 0.25000kg

 View more versions of this product

Product Code. 15866304

This item is currently unavailable or has been discontinued.
View the product page for possible alternatives.


Description

Description

  • For use with pumps that incorporate cassettes into the pumphead design.
  • Color-coded stops.
  • Inexpensive all-around tubing for general laboratory applications.
  • Transparent.
  • Resistant to almost all inorganic chemicals.
  • Smooth polished inner wall.
  • Low gas permeability.
  • Non-aging and non-oxidizing.
Specifications

Specifications

Orange-Green
Tygon LMT-55
General laboratory applications, pumps with cassettes in pumphead design
12/Pk
I.D. 0.38 mm
74°C
0.38 mm
400 mm
−50°C
Acids: Good, Alkaline solutions: Good, Solvents: Not rec., Pressure: Fair, Vacuum: Good, Viscous media: Excellent, Sterile media: Limited
Transparent
Product Suggestions

Product Suggestions

Videos
SDS
Documents

Documents

Certificates
Special Offers

Special Offers