missing translation for 'onlineSavingsMsg'
Learn More

(3’S,3R,4S)-Desfluoro Ezetimibe, TRC

Product Code. 30357601
Change view
Click to view available options
Quantity:
10 mg
100 mg
Unit Size:
100mg
10mg
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
30357601 10 mg 10mg
30359006 100 mg 100mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Product Code. 30357601 Supplier Toronto Research Chemicals Supplier No. TRCD28993510MG
 Restricted Item
Estimated Shipment: 14-08-2026
Add to basket
Add to basket
This item is not returnable. View return policy

High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 190595-66-5
Molecular Formula C24 H22 F N O3
Molecular Weight (g/mol) 391.43
Synonym (3R,4S)-1-(4-Fluorophenyl)-4-(4-hydroxyphenyl)-3-[(3S)-3-hydroxy-3-phenylpropyl]-2-azetidinone, 2-Azetidinone, 1-(4-fluorophenyl)-4-(4-hydroxyphenyl)-3-(3-hydroxy-3-phenylpropyl)-, [3R-[3α(S*),4β]]- (9CI)
IUPAC Name (3R,4S)-1-(4-fluorophenyl)-4-(4-hydroxyphenyl)-3-[(3S)-3-hydroxy-3-phenylpropyl]azetidin-2-one
SMILES O[C@@H](CC[C@@H]1[C@H](N(C1=O)c2ccc(F)cc2)c3ccc(O)cc3)c4ccccc4

Specifications

Quantity 10 mg
InChI Formula InChI=1S/C24H22FNO3/c25-18-8-10-19(11-9-18)26-23(17-6-12-20(27)13-7-17)21(24(26)29)14-15-22(28)16-4-2-1-3-5-16/h1-13,21-23,27-28H,14-15H2/t21-,22+,23-/m1/s1
Formula Weight 391.1584
Percent Purity >95%
Chemical Name or Material (3R,4S)-1-(4-Fluorophenyl)-4-(4-hydroxyphenyl)-3-[(3S)-3-hydroxy-3-phenylpropyl]-2-azetidinone
Grade HPLC
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.