missing translation for 'onlineSavingsMsg'
Learn More

3-Quinuclidinone Hydrochloride, TRC

Product Code. 30424811
Change view
Click to view available options
Quantity:
25 g
Unit Size:
25g
Product Code. Quantity unitSize
30424811 25 g 25g
1 options
This item is not returnable. View return policy

Product Code. 30424811

Brand: Toronto Research Chemicals TRCQ79579525G

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.

TRUSTED_SUSTAINABILITY

Specifications

Chemical Name or Material 3-Quinuclidinone Hydrochloride
CAS 1193-65-3
Molecular Formula C7H12ClNO
Quantity 25 g
Synonym 1-Azabicyclo[2.2.2]octan-3-one Hydrochloride, 3-Quinuclidinone Hydrochloride, 3-Oxoquinuclidine Hydrochloride
InChI Formula InChI=1S/C7H11NO.ClH/c9-7-5-8-3-1-6(7)2-4-8;/h6H,1-5H2;1H
SMILES Cl.O=C1CN2CCC1CC2
IUPAC Name 1-azabicyclo[2.2.2]octan-3-one;hydrochloride
Molecular Weight (g/mol) 161.63
Formula Weight 161.0607
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.