missing translation for 'onlineSavingsMsg'
Learn More

3-Quinuclidinone hydrochloride, 98+%, Thermo Scientific Chemicals

Product Code. 11450923
Change view
Click to view available options
Quantity:
5 g
25 g
100 g
Unit Size:
100g
25g
5g
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
11450923 25 g 25g
11440923 5 g 5g
11460923 100 g 100g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
This item is not returnable. View return policy
Product Code. 11450923 Supplier Thermo Scientific Alfa Aesar Supplier No. A13320.14
Request Bulk or Custom Formats

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 1193-65-3
Molecular Formula C7H12ClNO
Molecular Weight (g/mol) 161.629
MDL Number MFCD00137391
InChI Key RFDPHKHXPMDJJD-UHFFFAOYSA-N
Synonym 3-quinuclidinone hydrochloride, quinuclidin-3-one hydrochloride, 3-quinuclidone hydrochloride, 1-azabicyclo 2.2.2 octan-3-one hydrochloride, 3-quinuclidnone hydrochloride, 1-azabicyclo 2.2.2 octan-3-one, hydrochloride, 1-aza-3-oxobicyclo 2.2.2 octane, quinuclidin-3-one, chloride, 1-azabicyclo 2.2.2 octan-3-one, hydrochloride 1:1
PubChem CID 102019
IUPAC Name 1-azabicyclo[2.2.2]octan-3-one;hydrochloride
SMILES C1CN2CCC1C(=O)C2.Cl

Specifications

Melting Point ∼300°C (decomposition)
Quantity 25 g
Beilstein 3695039
Sensitivity Hygroscopic
Formula Weight 161.63
Percent Purity ≥98%
Chemical Name or Material 3-Quinuclidinone hydrochloride

RUO – Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.