Learn More
3-Quinuclidinol, 98+%, Thermo Scientific Chemicals
Description
3-Quinuclidinol acts as a chiral building block for various antimuscarinic agents and other active pharmaceutical ingredients. It serves as a catalyst for condensation of methyl vinyl ketone with aldehydes. It is also used as a reagent for cleavage of beta-keto and vinylogous beta-keto esters. Further, it plays an important role as a synthon in the preparation of cholinergic receptor ligands and anesthetics. In addition to this, it is used in chemoselective alfa-iodination of acrylic esters through Morita-Baylis-Hillman reaction.
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
3-Quinuclidinol acts as a chiral building block for various antimuscarinic agents and other active pharmaceutical ingredients. It serves as a catalyst for condensation of methyl vinyl ketone with aldehydes. It is also used as a reagent for cleavage of beta-keto and vinylogous beta-keto esters. Further, it plays an important role as a synthon in the preparation of cholinergic receptor ligands and anesthetics. In addition to this, it is used in chemoselective alfa-iodination of acrylic esters through Morita-Baylis-Hillman reaction.
Solubility
Soluble in water.
Notes
Incompatible with oxidizing agents.
Specifications
Melting Point | 217°C to 224°C |
pH | 10 to 12 |
Quantity | 100 g |
UN Number | UN3263 |
Beilstein | 104327 |
Sensitivity | Air Sensitive |
Merck Index | 14,8082 |
Solubility Information | Soluble in water. |
Formula Weight | 127.19 |
Percent Purity | ≥98% |
Show More |
RUO – Research Use Only