missing translation for 'onlineSavingsMsg'
Learn More

3-(quinoxalin-2-yl)prop-2-enoic Acid, TRC

Product Code. 30384331
Change view
Click to view available options
Quantity:
50 mg
Unit Size:
50mg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
30384331 50 mg 50mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Product Code. 30384331 Supplier Toronto Research Chemicals Supplier No. TRCE59244050MG

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.

TRUSTED_SUSTAINABILITY

Specifications

Chemical Name or Material 3-(quinoxalin-2-yl)prop-2-enoic Acid
CAS 1593-24-4
Molecular Formula C11 H8 N2 O2
Quantity 50 mg
Synonym 3-(2-Quinoxalinyl)-2-propenoic acid, NSC 158311
InChI Formula InChI=1S/C11H8N2O2/c14-11(15)6-5-8-7-12-9-3-1-2-4-10(9)13-8/h1-7H,(H,14,15)/b6-5+
SMILES OC(=O)\C=C\c1cnc2ccccc2n1
IUPAC Name (E)-3-quinoxalin-2-ylprop-2-enoic acid
Molecular Weight (g/mol) 200.193
Formula Weight 200.059
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.