missing translation for 'onlineSavingsMsg'
Learn More

3-Quinolineboronic Acid, TRC

Product Code. 30428292
Cambia vista
Click to view available options
Quantity:
2.5 g
Dimensione della confezione:
2.5g
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Unit Size
30428292 2.5 g 2.5g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Product Code. 30428292 Supplier Toronto Research Chemicals Supplier No. TRCQ7004502.5G
 Restricted Item
Estimated Shipment: 22-10-2026
Add to basket
Add to basket

missing translation for 'validMainframeMsg'

This item is not returnable. View return policy

High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.

TRUSTED_SUSTAINABILITY

Specifications

Chemical Name or Material 3-Quinolineboronic Acid
CAS 191162-39-7
Molecular Formula C9 H8 B N O2
Quantity 2.5 g
Synonym B-3-Quinolinylboronic Acid, 3-Quinolinylboronic Acid, Quinolin-3-ylboronic Acid
InChI Formula InChI=1S/C9H8BNO2/c12-10(13)8-5-7-3-1-2-4-9(7)11-6-8/h1-6,12-13H
SMILES OB(O)c1cnc2ccccc2c1
IUPAC Name quinolin-3-ylboronic acid
Molecular Weight (g/mol) 172.976
Formula Weight 173.065
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.