missing translation for 'onlineSavingsMsg'
Learn More

3-(quinolin-2-yl)propanoic Acid, TRC

Product Code. 30383125
Change view
Click to view available options
Quantity:
25 mg
50 mg
250 mg
Unit Size:
250mg
50mg
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
30383125 50 mg 50mg
30383930 25 mg -
30383213 250 mg 250mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
This item is not returnable. View return policy
Product Code. 30383125 Supplier Toronto Research Chemicals Supplier No. TRCE59244550MG

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 39111-94-9
Molecular Formula C12H11NO2
Molecular Weight (g/mol) 201.22
SMILES OC(=O)CCc1ccc2ccccc2n1

Specifications

Quantity 50 mg
InChI Formula InChI=1S/C12H11NO2/c14-12(15)8-7-10-6-5-9-3-1-2-4-11(9)13-10/h1-6H,7-8H2,(H,14,15)
Chemical Name or Material 3-(quinolin-2-yl)propanoic Acid
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.