missing translation for 'onlineSavingsMsg'
Learn More

3-Pyrrolidinol, 97%

Product Code. 10071254 Shop All Thermo Scientific Chemicals Products
Change view
Click to view available options
Quantity:
1 g
5 g
Packaging:
Glass bottle
Unit Size:
1g
5g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Packaging unitSize
10071254 1 g Glass bottle 1g
10536864 5 g Glass bottle 5g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Product Code. 10071254 Supplier Thermo Scientific Acros Supplier No. 425750010
Request Bulk or Custom Formats

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

CAS: 40499-83-0 | C4H9NO | 87.12 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 40499-83-0
Molecular Formula C4H9NO
Molecular Weight (g/mol) 87.12
MDL Number MFCD00005256
InChI Key JHHZLHWJQPUNKB-UHFFFAOYNA-N
Synonym 3-pyrrolidinol, 3-hydroxypyrrolidine, dl-3-pyrrolidinol, 3-hydroxy pyrrolidine, dl-3-hydroxypyrrolidine, rs-3-hydroxypyrrolidine, 3-hydroxypyrrolidinehydrochloride, pyrrolidin-3ol, pyrrolidin-3-o, pyrrolidine-3-ol
PubChem CID 98210
IUPAC Name pyrrolidin-3-ol
SMILES OC1CCNC1

Specifications

CAS Min % 96.0
CAS Max % 100.0
Color Colorless to Yellow
Density 1.0760g/mL
Boiling Point 179°C
Flash Point >110°C
Infrared Spectrum Authentic
Assay Percent Range 96% min. (GC)
Packaging Glass bottle
Refractive Index 1.4881
Quantity 1 g
Specific Gravity 1.076
Formula Weight 87.12
Percent Purity 97%
Physical Form Liquid
Chemical Name or Material 3-Pyrrolidinol
Show More Show Less
compliance-icons
Product Identifier
  • 3-Pyrrolidinol
Signal Word
  • Warning
Hazard Category
  • Serious eye damage/eye irritation Category 2
  • Skin corrosion/irritation Category 2
  • Specific target organ toxicity Category 3
Hazard Statement
  • H319-Causes serious eye irritation.
  • H335-May cause respiratory irritation.
  • H315-Causes skin irritation.
Precautionary Statement
  • P280-Wear protective gloves/protective clothing/eye protection/face protection.
  • P302+P352-IF ON SKIN: Wash with plenty of water/.
  • P304+P340-IF INHALED: Remove person to fresh air and keep comfortable for breathing.
  • P305+P351+P338-IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
  • P312-Call a POISON CENTER/doctor/if you feel unwell.

RUO – Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.