Learn More
(3-Pyrrolidin-1-ylphenyl)methylamine, ≥90%, Maybridge™
Brand: Maybridge CC38513CB
Code : 31
Additional Details : CAS Number : 175696-70-5 Weight : 0.05025kg Transport : UN number : 2735 Chem class : 8 Pack group : III
Packaging | Amber Glass Bottle |
---|---|
Quantity | 250mg |
Chemical Identifiers
175696-70-5 | |
176.263 | |
BXTWTBYGOKOWQX-UHFFFAOYSA-N | |
7162061 | |
C1CCN(C1)C2=CC=CC(=C2)CN |
C11H16N2 | |
MFCD07772827 | |
3-pyrrolidin-1-ylphenyl methylamine, 3-pyrrolidin-1-yl phenyl methanamine, 3-pyrrolidinobenzylamine, 1-3-pyrrolidin-1-yl phenyl methanamine, 3-pyrrolidin-1-ylphenyl methanamine, 3-pyrrolizinobenzylamine, 3-pyrrolidin-1-yl benzylamine, 3-pyrrolidinylphenyl methylamine, 3-pyrrolidin-1-yl phenyl methylamine | |
(3-pyrrolidin-1-ylphenyl)methanamine |
Specifications
(3-Pyrrolidin-1-ylphenyl)methylamine | |
250mg | |
89.5 | |
90% | |
MFCD07772827 | |
BXTWTBYGOKOWQX-UHFFFAOYSA-N | |
(3-pyrrolidin-1-ylphenyl)methanamine | |
7162061 | |
90% |
Amber Glass Bottle | |
175696-70-5 | |
100.0 | |
C11H16N2 | |
3-pyrrolidin-1-ylphenyl methylamine, 3-pyrrolidin-1-yl phenyl methanamine, 3-pyrrolidinobenzylamine, 1-3-pyrrolidin-1-yl phenyl methanamine, 3-pyrrolidin-1-ylphenyl methanamine, 3-pyrrolizinobenzylamine, 3-pyrrolidin-1-yl benzylamine, 3-pyrrolidinylphenyl methylamine, 3-pyrrolidin-1-yl phenyl methylamine | |
C1CCN(C1)C2=CC=CC(=C2)CN | |
176.263 | |
176.26 |