Learn More
(3-Pyrrolidin-1-ylphenyl)methanol, 97%, Thermo Scientific™
£26.55 - £101.00
Chemical Identifiers
CAS | 859850-72-9 |
---|---|
Molecular Formula | C11H15NO |
Molecular Weight (g/mol) | 177.247 |
MDL Number | MFCD07772826 |
InChI Key | MYIYSGIMXUQECR-UHFFFAOYSA-N |
Synonym | 3-pyrrolidin-1-ylphenyl methanol, 3-pyrrolidin-1-yl phenyl methanol, 3-1-pyrrolidinyl benzyl alcohol, 3-pyrrolidinylphenyl methan-1-ol, benzenemethanol,3-1-pyrrolidinyl |
PubChem CID | 7162062 |
IUPAC Name | (3-pyrrolidin-1-ylphenyl)methanol |
SMILES | C1CCN(C1)C2=CC=CC(=C2)CO |
Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
---|---|---|---|---|---|---|---|---|---|
Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
10045852
|
Thermo Scientific Maybridge
CC38509CB |
250 mg |
£26.55
250mg |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
10107772
|
Thermo Scientific Maybridge
CC38509DA |
1 g |
£101.00
1g |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
Description
Chemical Identifiers
859850-72-9 | |
177.247 | |
MYIYSGIMXUQECR-UHFFFAOYSA-N | |
7162062 | |
C1CCN(C1)C2=CC=CC(=C2)CO |
C11H15NO | |
MFCD07772826 | |
3-pyrrolidin-1-ylphenyl methanol, 3-pyrrolidin-1-yl phenyl methanol, 3-1-pyrrolidinyl benzyl alcohol, 3-pyrrolidinylphenyl methan-1-ol, benzenemethanol,3-1-pyrrolidinyl | |
(3-pyrrolidin-1-ylphenyl)methanol |
Specifications
859850-72-9 | |
100.0 | |
C11H15NO | |
3-pyrrolidin-1-ylphenyl methanol, 3-pyrrolidin-1-yl phenyl methanol, 3-1-pyrrolidinyl benzyl alcohol, 3-pyrrolidinylphenyl methan-1-ol, benzenemethanol,3-1-pyrrolidinyl | |
C1CCN(C1)C2=CC=CC(=C2)CO | |
177.247 | |
177.25 | |
(3-Pyrrolidin-1-ylphenyl)methanol |
96.5 | |
97% | |
MFCD07772826 | |
MYIYSGIMXUQECR-UHFFFAOYSA-N | |
(3-pyrrolidin-1-ylphenyl)methanol | |
7162062 | |
97% |
Safety and Handling
GHS H Statement:
Harmful if swallowed.
Causes skin irritation.
Harmful if inhaled.
Causes serious eye irritation.
May cause respiratory irritation.
GHS P Statement:
Avoid breathing dust/fume/gas/mist/vapors/spray.
IF SWALLOWED: Call a POISON CENTER or doctor/physician if you feel unwell.
IF INHALED: Remove victim to fresh air and keep at rest in a position comfortable for breathing.
IF ON SKIN: Wash with plenty of soap and water.
Wear protective gloves/protective clothing/eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
WARNING: The information provided on this web site was developed in compliance with European Union (EU) regulations and is correct to the best of our knowledge, information and belief at the date of its publication. The information given is designed only as a guide for safe handling and use. It is not to be considered as either a warranty or quality specification.
Warning