missing translation for 'onlineSavingsMsg'
Learn More

3-Pyrrolidin-1-ylbenzaldehyde, ≥95%, Thermo Scientific™

Product Code. 10779953
Click to view available options
Quantity:
250 mg
1 g
5 g
This item is not returnable. View return policy
This item is not returnable. View return policy

Chemical Identifiers

CAS 857283-89-7
Molecular Formula C11H13NO
Molecular Weight (g/mol) 175.23
MDL Number MFCD07772825
InChI Key KELYFFMWYRNGTF-UHFFFAOYSA-N
Synonym 3-pyrrolidin-1-yl benzaldehyde, 3-1-pyrrolidinyl benzaldehyde, benzaldehyde,3-1-pyrrolidinyl, 3-pyrrolidinylbenzaldehyde
PubChem CID 7164600
SMILES O=CC1=CC(=CC=C1)N1CCCC1

Specifications

CAS Min % 94.5
CAS Max % 100.0
Assay Percent Range 95%
Packaging Amber glass bottle
Quantity 1 g
IUPAC Name 3-(pyrrolidin-1-yl)benzaldehyde
Formula Weight 175.23
Percent Purity 95%
Chemical Name or Material 3-Pyrrolidin-1-ylbenzaldehyde
missing translation for 'provideContentCorrection'

missing translation for 'productTitle'