missing translation for 'onlineSavingsMsg'
Learn More

3-Pyrrolidin-1-ylaniline, 97%, Thermo Scientific™

Product Code. 10177812
Click to view available options
Quantity:
1 g
10 g
Packaging:
Amber Glass Bottle
This item is not returnable. View return policy
This item is not returnable. View return policy

Chemical Identifiers

CAS 115833-93-7
Molecular Formula C10H14N2
Molecular Weight (g/mol) 162.236
InChI Key DNPBFTQZONVQDX-UHFFFAOYSA-N
PubChem CID 1518808
IUPAC Name 3-pyrrolidin-1-ylaniline
SMILES C1CCN(C1)C2=CC(=CC=C2)N

Specifications

Assay Percent Range 97%
Packaging Amber Glass Bottle
Quantity 1 g
Synonym 3-pyrrolidin-1-yl aniline, 3-pyrrolidin-1-yl-phenylamine, 3-1-pyrrolidinyl aniline, benzenamine,3-1-pyrrolidinyl, benzenamine, 3-1-pyrrolidinyl, 3-pyrrolidinylphenylamine, 3-pyrrolizinoaniline, acmc-20e1pl, 1-3-aminophenyl pyrrolidine
Formula Weight 162.23
Percent Purity 97%
Chemical Name or Material 3-Pyrrolidin-1-ylaniline
missing translation for 'provideContentCorrection'

missing translation for 'productTitle'