missing translation for 'onlineSavingsMsg'
Learn More

3-Pyrimidin-5-ylbenzoic acid, ≥97%, Thermo Scientific™

Product Code. 10412723
Change view
Click to view available options
Quantity:
250 mg
1 g
5 g
Unit Size:
1g
250mg
5g
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
10412723 250 mg 250mg
10137662 1 g 1g
10377114 5 g 5g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
This item is not returnable. View return policy
Product Code. 10412723 Supplier Thermo Scientific Maybridge Supplier No. CC41701CB
Request Bulk or Custom Formats

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

Chemical Identifiers

CAS 852180-74-6
Molecular Formula C11H8N2O2
Molecular Weight (g/mol) 200.197
MDL Number MFCD05864810
InChI Key URQRFVRGUHUTMI-UHFFFAOYSA-N
Synonym 3-pyrimidin-5-yl-benzoic acid, 3-pyrimidin-5-yl benzoic acid, 3-5-pyrimidinyl benzoic acid, benzoic acid,3-5-pyrimidinyl, benzoic acid, 3-5-pyrimidinyl, 3-pyrimidin-5-yl-benzoicacid, benzoic acid, 3-5-pyrimidinyl-9ci
PubChem CID 4187250
IUPAC Name 3-pyrimidin-5-ylbenzoic acid
SMILES C1=CC(=CC(=C1)C(=O)O)C2=CN=CN=C2

Specifications

CAS Min % 96.5
CAS Max % 100.0
Assay Percent Range 97%
Packaging Amber glass bottle
Quantity 250 mg
Formula Weight 200.2
Percent Purity 97%
Chemical Name or Material 3-Pyrimidin-5-ylbenzoic acid
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.