missing translation for 'onlineSavingsMsg'
Learn More

3-Pyrimidin-5-ylaniline, ≥97%, Thermo Scientific™

Product Code. 10164192
Change view
Click to view available options
Quantity:
250 mg
1 g
5 g
Unit Size:
1g
250mg
5g
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
10164192 1 g 1g
10558253 250 mg 250mg
10011844 5 g 5g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
This item is not returnable. View return policy
Product Code. 10164192 Supplier Thermo Scientific Maybridge Supplier No. CC41714DA
Request Bulk or Custom Formats

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

Chemical Identifiers

CAS 69491-59-4
Molecular Formula C10H9N3
Molecular Weight (g/mol) 171.203
MDL Number MFCD06802522
InChI Key DZEIKJMNXHOFHL-UHFFFAOYSA-N
PubChem CID 7162049
IUPAC Name 3-pyrimidin-5-ylaniline
SMILES C1=CC(=CC(=C1)N)C2=CN=CN=C2

Specifications

CAS Min % 96.5
CAS Max % 100.0
Assay Percent Range 97%
Packaging Amber glass bottle
Quantity 1 g
Synonym 3-pyrimidin-5-yl aniline, 3-5-pyrimidinyl aniline, benzenamine,3-5-pyrimidinyl, 3-5-pyrimidyl aniline, 3-5-pyrimidyl-aniline, 3-pyrimidin-5-ylphenylamine, 3-pyrimidin-5-yl-phenylamine, 3-pyrimidin-5-yl phenylamine, benzenamine, 3-5-pyrimidinyl
Formula Weight 171.2
Percent Purity 97%
Chemical Name or Material 3-Pyrimidin-5-ylaniline
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.