missing translation for 'onlineSavingsMsg'
Learn More

3-Pyrimidin-2-ylbenzylamine, 97%, Thermo Scientific™

Product Code. 10275324
Change view
Click to view available options
Quantity:
250 mg
1 g
Packaging:
Amber Glass Bottle
Unit Size:
1g
250mg
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Packaging unitSize
10275324 1 g Amber Glass Bottle 1g
10472454 250 mg Amber Glass Bottle 250mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Product Code. 10275324 Supplier Thermo Scientific Maybridge Supplier No. CC56313DA
Request Bulk or Custom Formats

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

Chemical Identifiers

CAS 910036-92-9
Molecular Formula C11H11N3
Molecular Weight (g/mol) 185.23
MDL Number MFCD09702384
InChI Key MXSXDKVANPZCBQ-UHFFFAOYSA-N
Synonym 3-pyrimidin-2-ylbenzylamine, 3-pyrimidin-2-yl phenyl methanamine, 3-pyrimidin-2-yl benzylamine, 1-3-pyrimidin-2-yl phenyl methanamine, 3-pyrimidin-2-ylphenyl methylamine, benzenemethanamine, 3-2-pyrimidinyl
PubChem CID 24229566
IUPAC Name (3-pyrimidin-2-ylphenyl)methanamine
SMILES NCC1=CC=CC(=C1)C1=NC=CC=N1

Specifications

CAS Min % 94.5
CAS Max % 100.0
Assay Percent Range 95%
Packaging Amber Glass Bottle
Quantity 1 g
Formula Weight 185.23
Percent Purity 95%
Chemical Name or Material 3-Pyrimidin-2-ylbenzylamine
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.