missing translation for 'onlineSavingsMsg'
Learn More

3-Pyridylacetonitrile, 98%

Product Code. 10747855 Shop All Thermo Scientific Chemicals Products
Change view
Click to view available options
Quantity:
1 g
5 g
25 g
Packaging:
Glass bottle
Unit Size:
1g
25g
5g
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Packaging unitSize
10747855 25 g Glass bottle 25g
10153064 1 g Glass bottle 1g
10326414 5 g Glass bottle 5g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
This item is not returnable. View return policy
Product Code. 10747855 Supplier Thermo Scientific Acros Supplier No. 131970250
Request Bulk or Custom Formats

Please to purchase this item. Need a web account? Register with us today!

This item is currently unavailable or has been discontinued.
View the product page for possible alternatives.
View alternative products

This item is not returnable. View return policy

CAS: 6443-85-2 | C7H6N2 | 118.14 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 6443-85-2
Molecular Formula C7H6N2
Molecular Weight (g/mol) 118.14
MDL Number MFCD00006406
InChI Key OIPHWUPMXHQWLR-UHFFFAOYSA-N
Synonym 3-pyridineacetonitrile, 3-pyridylacetonitrile, 2-pyridin-3-yl acetonitrile, pyridine-3-acetonitrile, 3-cyanomethylpyridine, 3-cyanomethyl pyridine, pyridin-3-ylacetonitrile, 2-3-pyridyl ethanenitrile, pyridin-3-yl-acetonitrile, 3-pyridyl acetonitrile
PubChem CID 80923
IUPAC Name 2-pyridin-3-ylacetonitrile
SMILES N#CCC1=CC=CN=C1

Specifications

CAS Min % 97.5
CAS Max % 100.0
Color Colorless to Yellow
Density 1.1000g/mL
Boiling Point 105°C to 108°C (1.5 mmHg)
Flash Point 98°C
Infrared Spectrum Authentic
Assay Percent Range 97.5 % min. (GC)
Packaging Glass bottle
Linear Formula (C5H4N)CH2C∽N
Refractive Index 1.5289
Quantity 25 g
Specific Gravity 1.1
Formula Weight 118.14
Percent Purity 98%
Physical Form Liquid
Chemical Name or Material 3-Pyridylacetonitrile
Show More Show Less
compliance-icons
Product Identifier
  • 3-Pyridylacetonitrile
Signal Word
  • Warning
Hazard Category
  • Acute toxicity Category 4
  • Serious eye damage/eye irritation Category 2
  • Skin corrosion/irritation Category 2
  • Specific target organ toxicity Category 3
Hazard Statement
  • H315-Causes skin irritation.
  • H319-Causes serious eye irritation.
  • H335-May cause respiratory irritation.
  • H312-Harmful in contact with skin.
  • H332-Harmful if inhaled.
  • H302-Harmful if swallowed.
Precautionary Statement
  • P261-Avoid breathing dust/fume/gas/mist/vapours/spray.
  • P280-Wear protective gloves/protective clothing/eye protection/face protection.
  • P301+P312-IF SWALLOWED: Call a POISON CENTER or doctor/physician/ if you feel unwell.
  • P302+P352-IF ON SKIN: Wash with plenty of water/.
  • P304+P340-IF INHALED: Remove person to fresh air and keep comfortable for breathing.
  • P305+P351+P338-IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.

RUO – Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.