missing translation for 'onlineSavingsMsg'
Learn More

3-Pyridylacetic Acid Hydrochloride, TRC

Product Code. 17973595
Change view
Click to view available options
Quantity:
1 g
Unit Size:
1g
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Unit Size
17973595 1 g 1g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Product Code. 17973595 Supplier Toronto Research Chemicals Supplier No. TRCP9917651G
 Restricted Item
Estimated Shipment: 11-09-2026
Add to basket
Add to basket
This item is not returnable. View return policy

High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.

TRUSTED_SUSTAINABILITY

Specifications

CAS 6419-36-9
Molecular Formula C7 H7 N O2 . Cl H
Quantity 1 g
Synonym 3-Pyridineacetic acid hydrochloride (1:1), 3-Pyridineacetic acid, hydrochloride (8CI,9CI), 2-(Pyridin-3-yl)acetic acid hydrochloride, 3-Pyridylacetic acid hydrochloride, Pyridin-3-ylacetic acid hydrochloride
InChI Formula InChI=1S/C7H7NO2.ClH/c9-7(10)4-6-2-1-3-8-5-6;/h1-3,5H,4H2,(H,9,10);1H
SMILES Cl.OC(=O)Cc1cccnc1
IUPAC Name 2-pyridin-3-ylacetic acid;hydrochloride
Molecular Weight (g/mol) 173.6
Formula Weight 173.0244
Chemical Name or Material 3-Pyridylacetic acid hydrochloride
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.