missing translation for 'onlineSavingsMsg'
Learn More

3-Pyridinesulfonic acid, 98%

Product Code. 10173660 Shop All Thermo Scientific Chemicals Products
Change view
Click to view available options
Quantity:
100 g
500 g
Packaging:
Plastic bottle
Unit Size:
100g
500g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Packaging unitSize
10173660 500 g Plastic bottle 500g
10163660 100 g Plastic bottle 100g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Product Code. 10173660 Supplier Thermo Scientific Acros Supplier No. 131925000
Request Bulk or Custom Formats

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

CAS: 636-73-7 | C5H4NO3S | 158.15 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 636-73-7
Molecular Formula C5H4NO3S
Molecular Weight (g/mol) 158.15
MDL Number MFCD00006381
InChI Key DVECLMOWYVDJRM-UHFFFAOYSA-M
Synonym 3-pyridinesulfonic acid, pyridine-3-sulphonic acid, 3-pyridylsulfonic acid, 3-pyridine sulfonic acid, 3-sulfopyridine, beta-sulfopyridine, 3-pyridinesulfonicacid, pubchem9608, pyridin-3-sulfonic acid, acmc-209nh7
PubChem CID 69468
IUPAC Name pyridine-3-sulfonic acid
SMILES [O-]S(=O)(=O)C1=CC=CN=C1

Specifications

CAS Min % 97.5
CAS Max % 100.0
Melting Point >300.0°C
Color White
Flash Point >66°C
Infrared Spectrum Authentic
Assay Percent Range 98%
Packaging Plastic bottle
Linear Formula (C5H4N)SO3H
Quantity 500 g
Beilstein 22, 387
Solubility Information (5% in water) Clear colorless
Formula Weight 159.16
Percent Purity 98%
Physical Form Crystalline Powder
Chemical Name or Material 3-Pyridinesulfonic acid, 98%
Show More Show Less
compliance-icons
Product Identifier
  • 3-Pyridinesulfonic acid
Signal Word
  • Danger
Hazard Category
  • Serious eye damage/eye irritation Category 1
  • Skin corrosion/irritation Category 1 B
Hazard Statement
  • H318-Causes serious eye damage.
  • H314-Causes severe skin burns and eye damage.
Precautionary Statement
  • P280-Wear protective gloves/protective clothing/eye protection/face protection.
  • P301+P312-IF SWALLOWED: Call a POISON CENTER or doctor/physician/ if you feel unwell.
  • P301+P330+P331-IF SWALLOWED: rinse mouth. Do NOT induce vomiting.
  • P305+P351+P338-IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
  • P310-Immediately call a POISON CENTER/doctor/.

RUO – Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.