missing translation for 'onlineSavingsMsg'
Learn More

3-Pyridinecarboxaldehyde, 98%

Product Code. p-4509898 Shop All Thermo Scientific Chemicals Products
Click to view available options
Quantity:
25 g
100 g
500 g
Packaging:
Glass bottle
Unit Size:
100g
25g
500g
This item is not returnable. View return policy

Product Code. 10143470

Brand: Thermo Scientific Acros 131830250

Request Bulk or Custom Formats

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

CAS: 500-22-1 | C6H5NO | 107.11 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 500-22-1
Molecular Formula C6H5NO
Molecular Weight (g/mol) 107.11
InChI Key QJZUKDFHGGYHMC-UHFFFAOYSA-N
Synonym 3-pyridinecarboxaldehyde, nicotinaldehyde, 3-pyridinaldehyde, pyridine-3-carboxaldehyde, 3-pyridylcarboxaldehyde, 3-pyridylaldehyde, nicotinealdehyde, 3-formylpyridine, pyridine-3-aldehyde, nicotinic aldehyde
PubChem CID 10371
ChEBI CHEBI:28345
IUPAC Name pyridine-3-carbaldehyde
SMILES C1=CC(=CN=C1)C=O

Specifications

Color Yellow to Brown
Density 1.1420g/mL
Boiling Point 95.0°C to 97.0°C (15.0 mmHg)
Flash Point 84°C
Infrared Spectrum Authentic
Assay Percent Range 97.5% min. (GC)
Packaging Glass bottle
Linear Formula (C5H4N)COH
Refractive Index 1.5480 to 1.5500
Quantity 25 g
Beilstein 21,I,288
Specific Gravity 1.142
Solubility Information Solubility in water: miscible. Other solubilities: soluble in most organic solvents
Formula Weight 107.11
Percent Purity 98%
Physical Form Liquid
Chemical Name or Material 3-Pyridinecarboxaldehyde
Show More Show Less
compliance-icons compliance-icons compliance-icons
Product Identifier
  • 3-Pyridinecarboxaldehyde
Signal Word
  • Danger
Hazard Category
  • Acute toxicity Category 4
  • LONG-TERM AQUATIC HAZARD Chronic 3
  • Serious eye damage/eye irritation Category 1
  • Skin corrosion/irritation Category 2
  • Skin sensitiser Category 1
  • Specific target organ toxicity Category 3
Hazard Statement
  • H226-Flammable liquid and vapour.
  • H302-Harmful if swallowed.
  • H315-Causes skin irritation.
  • H317-May cause an allergic skin reaction.
  • H318-Causes serious eye damage.
  • H335-May cause respiratory irritation.
  • H412-Harmful to aquatic life with long lasting effects.
Precautionary Statement
  • P210-Keep away from heat, hot surfaces, sparks, open flames and other ignition sources. No smoking.
  • P280-Wear protective gloves/protective clothing/eye protection/face protection.
  • P301+P330+P331-IF SWALLOWED: rinse mouth. Do NOT induce vomiting.
  • P303+P361+P353-IF ON SKIN (or hair): Take off immediately all contaminated clothing. Rinse skin with water/ shower.
  • P304+P340-IF INHALED: Remove person to fresh air and keep comfortable for breathing.
  • P305+P351+P338-IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
  • P310-Immediately call a POISON CENTER/doctor/.

RUO – Research Use Only

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.