missing translation for 'onlineSavingsMsg'
Learn More

3-Pyridinealdoxime, 98%, ACROS Organics™

Product Code. 10295370 Shop All Thermo Scientific Chemicals Products
Change view
Click to view available options
Quantity:
5 g
25 g
Packaging:
Glass bottle
Unit Size:
25g
5g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Packaging unitSize
10295370 25 g Glass bottle 25g
10068541 5 g Glass bottle 5g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Product Code. 10295370 Supplier Thermo Scientific Acros Supplier No. 131800250
Request Bulk or Custom Formats

Please to purchase this item. Need a web account? Register with us today!

This item is currently unavailable or has been discontinued.
View the product page for possible alternatives.
View alternative products

This item is not returnable. View return policy

Chemical Identifiers

CAS 1193-92-6
Molecular Formula C6H6N2O
Molecular Weight (g/mol) 122.13
MDL Number MFCD00006409,MFCD00006409
InChI Key YBKOPFQCLSPTPV-VMPITWQZSA-N
Synonym nicotinaldoxime, 3-pyridinealdoxime, nicotinaldehyde oxime, pyridine-3-aldoxime, 3-pyridinecarboxaldehyde, oxime, nicotinaldehyde, oxime, 3-pyridine-aldoxime, pyridine, 3-formyl-, oxime, hydroxyimino-3-pyridylmethane, pyridinaldoxim
PubChem CID 5371219
IUPAC Name (E)-N-[(pyridin-3-yl)methylidene]hydroxylamine
SMILES O\N=C\C1=CC=CN=C1

Specifications

CAS Min % 97.5
CAS Max % 100.0
Melting Point 149°C to 153°C
Color White
Infrared Spectrum Authentic
Assay Percent Range 98%
Packaging Glass bottle
Quantity 25 g
Formula Weight 122.13
Percent Purity ≥97.5%
Physical Form Fine Crystalline Powder
Chemical Name or Material 3-Pyridinealdoxime, 98%
Show More Show Less
compliance-icons
Product Identifier
  • 3-Pyridinealdoxime
Signal Word
  • Warning
Hazard Category
  • Acute toxicity Category 4
  • Serious eye damage/eye irritation Category 2
  • Skin corrosion/irritation Category 2
  • Specific target organ toxicity Category 3
Hazard Statement
  • H315-Causes skin irritation.
  • H319-Causes serious eye irritation.
  • H335-May cause respiratory irritation.
  • H332-Harmful if inhaled.
  • H302-Harmful if swallowed.
Precautionary Statement
  • P261-Avoid breathing dust/fume/gas/mist/vapours/spray.
  • P280-Wear protective gloves/protective clothing/eye protection/face protection.
  • P301+P312-IF SWALLOWED: Call a POISON CENTER or doctor/physician/ if you feel unwell.
  • P302+P352-IF ON SKIN: Wash with plenty of water/.
  • P304+P340-IF INHALED: Remove person to fresh air and keep comfortable for breathing.
  • P305+P351+P338-IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.