missing translation for 'onlineSavingsMsg'
Learn More

3-Pyridineacetonitrile, 98%, Thermo Scientific™

Product Code. 11421517
Change view
Click to view available options
Quantity:
1 g
5 g
25 g
Unit Size:
1g
25g
5g
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
11421517 1 g 1g
11431517 5 g 5g
11441517 25 g 25g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
This item is not returnable. View return policy
Product Code. 11421517 Supplier Thermo Scientific Alfa Aesar Supplier No. B24964.03
Request Bulk or Custom Formats

Please to purchase this item. Need a web account? Register with us today!

This item is currently unavailable or has been discontinued.
View the product page for possible alternatives.
View alternative products

This item is not returnable. View return policy

This Thermo Scientific brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific.

Applications
3-Pyridineacetonitrile can react with thiophene-2-carbaldehyde to get 2-pyridin-3-yl-3-thiophen-2-yl-acrylonitrile. It is also used as pharmaceutical intermediates.

Solubility
Soluble in alcohol, ether, ketone. Insoluble in water.

Notes
Store in cool place. Keep container tightly closed in a dry and well-ventilated place. Incompatible materials are oxidizing agents. Store at 4°C.
TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 6443-85-2
Molecular Formula C7H6N2
Molecular Weight (g/mol) 118.14
MDL Number MFCD00006406
InChI Key OIPHWUPMXHQWLR-UHFFFAOYSA-N
Synonym 3-pyridineacetonitrile, 3-pyridylacetonitrile, 2-pyridin-3-yl acetonitrile, pyridine-3-acetonitrile, 3-cyanomethylpyridine, 3-cyanomethyl pyridine, pyridin-3-ylacetonitrile, 2-3-pyridyl ethanenitrile, pyridin-3-yl-acetonitrile, 3-pyridyl acetonitrile
PubChem CID 80923
SMILES N#CCC1=CC=CN=C1

Specifications

Density 1.108
Quantity 1 g
UN Number UN3276
Solubility Information Soluble in alcohol,ether,ketone. Insoluble in water.
IUPAC Name 2-pyridin-3-ylacetonitrile
Formula Weight 118.14
Percent Purity 98%
Chemical Name or Material 3-Pyridineacetonitrile
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.