missing translation for 'onlineSavingsMsg'
Learn More

3-Pyrid-4-ylbenzoic acid, 97%, Thermo Scientific™

Product Code. 10750054
Change view
Click to view available options
Quantity:
250 mg
1 g
5 g
Unit Size:
1g
250mg
5g
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
10750054 250 mg 250mg
10145632 1 g 1g
10337254 5 g 5g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
This item is not returnable. View return policy
Product Code. 10750054 Supplier Thermo Scientific Maybridge Supplier No. CC35601CB
Request Bulk or Custom Formats

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

Chemical Identifiers

CAS 4385-78-8
Molecular Formula C12H9NO2
Molecular Weight (g/mol) 199.209
InChI Key IYGIZNZSONLPSI-UHFFFAOYSA-N
Synonym 3-pyridin-4-yl benzoic acid, 3-pyridin-4-yl-benzoic acid, 3-pyrid-4-ylbenzoic acid, 3-4-pyridyl benzoic acid, 4-3-carboxyphenyl pyridine, 3-4-pyridinyl benzoic acid, 3-pyridin-4-yl benzoic acid, benzoic acid,3-4-pyridinyl, 5fl
PubChem CID 2795562
IUPAC Name 3-pyridin-4-ylbenzoic acid
SMILES C1=CC(=CC(=C1)C(=O)O)C2=CC=NC=C2

Specifications

Assay Percent Range 95%
Quantity 250 mg
Formula Weight 199.21
Percent Purity 97%
Chemical Name or Material 3-Pyrid-4-ylbenzoic acid
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.