missing translation for 'onlineSavingsMsg'
Learn More

3-(Pyrid-2-yloxy)benzaldehyde, 97%, Thermo Scientific™

Product Code. 10124014
Change view
Click to view available options
Quantity:
250 mg
1 g
Packaging:
Amber Glass Bottle
Unit Size:
1g
250mg
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Packaging unitSize
10124014 250 mg Amber Glass Bottle 250mg
10764964 1 g Amber Glass Bottle 1g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Product Code. 10124014 Supplier Thermo Scientific Maybridge Supplier No. CC50204CB
Request Bulk or Custom Formats

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

Chemical Identifiers

CAS 137386-78-8
Molecular Formula C12H9NO2
Molecular Weight (g/mol) 199.209
MDL Number MFCD03840110
InChI Key YGXYNKLOBXMTGL-UHFFFAOYSA-N
Synonym 3-pyridin-2-yloxy benzaldehyde, 3-pyrid-2-yloxy benzaldehyde, 3-2-pyridinyloxy-benzaldehyde, 3-2-pyridyloxy benzaldehyde, 3-2-pyridinyloxy benzaldehyde
PubChem CID 2735363
IUPAC Name 3-pyridin-2-yloxybenzaldehyde
SMILES C1=CC=NC(=C1)OC2=CC=CC(=C2)C=O

Specifications

CAS Min % 96.5
CAS Max % 100.0
Assay Percent Range 97%
Packaging Amber Glass Bottle
Quantity 250 mg
Formula Weight 199.21
Percent Purity 97%
Chemical Name or Material 3-(Pyrid-2-yloxy)benzaldehyde
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.