missing translation for 'onlineSavingsMsg'
Learn More

3-Pyrid-2-ylbenzaldehyde, ≥97%, Thermo Scientific™

Product Code. 10780904
Change view
Click to view available options
Quantity:
250 mg
1 g
25 g
Unit Size:
1g
250mg
25g
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
10780904 250 mg 250mg
10177612 1 g 1g
10133914 25 g 25g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
This item is not returnable. View return policy
Product Code. 10780904 Supplier Thermo Scientific Maybridge Supplier No. CC41604CB
Request Bulk or Custom Formats

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

Chemical Identifiers

CAS 85553-53-3
Molecular Formula C12H9NO
Molecular Weight (g/mol) 183.21
MDL Number MFCD02684103
InChI Key SAPNGHSAYQXRPG-UHFFFAOYSA-N
Synonym 3-2-pyridyl benzaldehyde, 3-pyridin-2-yl benzaldehyde, 3-2-pyridinyl benzaldehyde, 3-pyridin-2-yl-benzaldehyde, 3-pyrid-2-ylbenzaldehyde, benzaldehyde, 3-2-pyridinyl, 2-3-formylphenyl pyridine, acmc-209q6q, benzaldehyde,3-2-pyridinyl, 5-2-pyridyl benzaldehyde
PubChem CID 3710039
IUPAC Name 3-pyridin-2-ylbenzaldehyde
SMILES C1=CC=NC(=C1)C2=CC(=CC=C2)C=O

Specifications

CAS Min % 96.5
CAS Max % 100.0
Assay Percent Range 97%
Packaging Amber Glass Bottle
Quantity 250 mg
Formula Weight 183.21
Percent Purity 97%
Chemical Name or Material 3-Pyrid-2-ylbenzaldehyde
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.