missing translation for 'onlineSavingsMsg'
Learn More

3-Piperidinemethanol, 96%

Product Code. 11336866
Change view
Click to view available options
Quantity:
5 g
25 g
Unit Size:
25g
5g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
11336866 5 g 5g
11346866 25 g 25g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Product Code. 11336866 Supplier Thermo Scientific Alfa Aesar Supplier No. L04264.06
Request Bulk or Custom Formats

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

CAS: 4606-65-9 | C6H13NO | 115.176 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 4606-65-9
Molecular Formula C6H13NO
Molecular Weight (g/mol) 115.176
MDL Number MFCD00005997
InChI Key VUNPWIPIOOMCPT-UHFFFAOYSA-N
Synonym 3-piperidinemethanol, 3-hydroxymethyl piperidine, piperidin-3-yl-methanol, 3-hydroxymethylpiperidine, 3-piperidinylmethanol, piperidine-3-methanol, 3-piperidine methanol, 3-piperidylmethanol, 3-piperidinyl methanol, piperidin-3-yl methanol
PubChem CID 107308
IUPAC Name piperidin-3-ylmethanol
SMILES C1CC(CNC1)CO

Specifications

Melting Point 58°C to 62°C
Density 1.025
Boiling Point 236°C
Flash Point >110°C (230°F)
Refractive Index 1.494
Quantity 5 g
UN Number UN3259
Beilstein 103058
Sensitivity Hygroscopic
Formula Weight 115.18
Percent Purity 96%
Chemical Name or Material 3-Piperidinemethanol
Show More Show Less

RUO – Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.