Learn More
Thermo Scientific Maybridge 3-Piperidinecarboxylic acid, 10g, 97%
Brand: Thermo Scientific Maybridge AC19919EA
3-Piperidinecarboxylic acid, 10g, 97%
Code : 31
Additional Details : CAS Number : 498-95-3 Weight : 0.06000kg
Brand: Thermo Scientific Maybridge AC19919EA
3-Piperidinecarboxylic acid, 10g, 97%
Code : 31
Additional Details : CAS Number : 498-95-3 Weight : 0.06000kg